Learn More
Biotium GloMelt™ Thermal Shift Protein Stability kit
Catalog No. NC1540596
Encompass_Preferred
GloMelt™ dye undergoes fluorescence enhancement upon binding to hydrophobic regions of denatured proteins. GloMelt™ can therefore be used to detect protein unfolding or protein thermal stability by performing a thermal shift assay, also called differential scanning fluorimetry or Thermofluor assay.
Catalog No. NC1540596 Supplier Biotium Supplier No. 33021T
GloMelt™ dye undergoes fluorescence enhancement upon binding to hydrophobic regions of denatured proteins. GloMelt™ can therefore be used to detect protein unfolding or protein thermal stability by performing a thermal shift assay, also called differential scanning fluorimetry or Thermofluor assay.