Learn More
Medchemexpress LLC HY-N2498 5mg Medchemexpress, Glomeratose A CAS:202471-84-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN24985MG
Encompass_Preferred
Medchemexpress, HY-N2498 5mg Glomeratose A CAS:202471-84-9 Glomeratose A is a lactate dehydrogenase inhibitor, isolated from Polygala tenuifolia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0400