Learn More
Technitrile GLOVE 12IN NITRILE 1000/C LRG
SDP
Catalog No. NC9208980
Encompass_Preferred
Glove, Nitrile, Powder Free, Style: Ambidextrous, Length: 12 in., Thickness: 6Mil, Grip Surface: Microtextured, cuff: Beaded, Tensile Strength: 12.9 MPa Minimum, Size: L, Chemical Resistance
This item is not returnable. View return policy
Catalog No. NC9208980 Supplier Technitrile Supplier No. TNT1203PFB
Glove, Nitrile, Powder Free, Style: Ambidextrous, Length: 12 in., Thickness: 6Mil, Grip Surface: Microtextured, cuff: Beaded, Tensile Strength: 12.9 MPa Minimum, Size: L, Chemical Resistance