Learn More
Technitrile GLOVE 12IN NITRILE 1000/C SM
SDP
Catalog No. NC9208977
Encompass_Preferred
Glove, Nitrile; Class 10 (M2.5) Powder Free; Small; Ambidextrous;12L x 0.006 in. T; Grip Surface Microtexture;Cuff Beaded; Tensile Strength 12.9MPa min; For critical environments;
This item is not returnable. View return policy
Catalog No. NC9208977 Supplier Technitrile Supplier No. TNT1201PFB
Glove, Nitrile; Class 10 (M2.5) Powder Free; Small; Ambidextrous;12L x 0.006 in. T; Grip Surface Microtexture;Cuff Beaded; Tensile Strength 12.9MPa min; For critical environments;