Learn More
Glas Col LLC Glove Bag
SDP
Supplier:  Glas Col LLC 108D X2717HEVAL
Encompass_Preferred
Glovebag 27" x 17" x 12" with 22" equipment opening in the front between two integral turned in gloves.Made of 3mil low density polyelthylene. With Handy Lok closure. One for evaluation purposes.
Catalog No. NC0424681