Learn More
Glas Col LLC Glove Bag
SDP
Supplier:  Glas Col LLC 108D X2717HG
Encompass_Preferred
Glove bag 27" x 17" x 12" with one 22" equipment opening in the front between two integral turned in gloves. Made of 3mil low density polyethelene. With HandyGlide closure.
Catalog No. NC0927249