Learn More
Glas Col LLC Glove Bag
SDP
Supplier:  Glas Col LLC 108D X2717H
Encompass_Preferred
Self contained chamber with one opening in the front between two integral gloves. Great when working with air or moisture sensitive materials. 27" x 17" x 12. Equipemnt sleeve 22".
Catalog No. NC9793587