Learn More
Glas Col LLC Glove Bag
SDP
Supplier:  Glas Col LLC 108D X3727H
Encompass_Preferred
Self contained chamber with one opening in the front between two integral gloves. Great when working with air or moisture sensitive materials. 37" x 27" x18". Equipment sleeve 33 1/2".
Catalog No. NC9841911