Learn More
Cole-Parmer Glove Bag 108DX-37-37H Inflatable Glove Chamber, 34.5" Equipment Sleeve, 6/pk
Catalog No. NC9336151 Shop All Cole-Parmer Products
Encompass_Preferred
An alternative to standard glove boxes—these durable, polyethylene glove chambers are reusable, yet priced to be disposable. Chambers are easy to use—just slide your equipment through the sleeve(s), seal, connect the gas port (accepts 3⁄8'' ID tubing) to your gas supply, and inflate. Glove bags have a 2.5 mil thickness and are made of polyethylene. Glove bags are not autoclaveable, nor are they sterile.
Catalog No. NC9336151 Supplier Cole-Parmer Supplier No. 0440838
An alternative to standard glove boxes—these durable, polyethylene glove chambers are reusable, yet priced to be disposable. Chambers are easy to use—just slide your equipment through the sleeve(s), seal, connect the gas port (accepts 3⁄8'' ID tubing) to your gas supply, and inflate. Glove bags have a 2.5 mil thickness and are made of polyethylene. Glove bags are not autoclaveable, nor are they sterile.