Learn More
Cole-Parmer Inflatable Glove Chamber, 22.5" Equipment Sleeve, 6/pk
Supplier:  Cole-Parmer 0440834
Encompass_Preferred
Glove Bag 108DX-27-27H Inflatable Glove Chamber, 22.5" Equipment Sleeve, 6/pk, Protect yourself and your materials quickly without giving up lab spaceEffectively eliminate oxygen and moisture during material manipulation operationsEstablish high-purity working atmosphere in minutesDisposable chambers simplify cleanup
Catalog No. NC0864960