Learn More
Grainger HONEYWELL NORTH Glove Box Gloves, 8 in For
Catalog No. NC2178845
Encompass_Preferred
The Honeywell North 8" Glove Box Gloves are made of natural rubber latex with a textured grip surface. They feature a straight cuff, 0.014" thickness, and are ambidextrous for use on either hand. These industrial gloves provide dexterity for handling small parts and objects while protecting hands from cuts, abrasions, oils, and greases. They can withstand temperatures from -20°F to 180°F. The gloves are sold individually and available in sizes small through x-large.
Catalog No. NC2178845 Supplier Grainger Supplier No. 4T480
The Honeywell North 8" Glove Box Gloves are made of natural rubber latex with a textured grip surface. They feature a straight cuff, 0.014" thickness, and are ambidextrous for use on either hand. These industrial gloves provide dexterity for handling small parts and objects while protecting hands from cuts, abrasions, oils, and greases. They can withstand temperatures from -20°F to 180°F. The gloves are sold individually and available in sizes small through x-large.