Learn More
Poltex Glove Box Holder with Bin
SDP
Catalog No. 22000730
Change view
Click to view available options
Mounting:
Magnetic Mount
VHB Tape
Wall Mount
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
22-000-730 Wall Mount
22-000-728 Magnetic Mount
22-000-729 VHB Tape
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-000-730 Supplier Poltex Supplier No. GB2BINW
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Break-resistant PETG glove box holder that holds 2 boxes of gloves, wipe tub, spray cleaner, hand sanitizer, etc.
  • Rubber bumpons on base for non-slip placement on countertop.
  • Choose from wall mount/countertop placement (screws/anchors included), magnet mount (powerful magnets included), or tape mount (VHB tape included).
Specifications
Product Type Glove Box Holder
Color Clear
Holds Disposable Glove Boxes
Material PETG
Mounting Wall Mount
No. of Compartments 2
Capacity 2 Boxes