Learn More
Cole-Parmer Plas-Labs 800-IRIS Glove Box Iris Diaphragm, 6" Diameter; 1 Pair
Catalog No. NC1564217 Shop All Cole-Parmer Products
Encompass_Preferred
Must be ordered when glove box is ordered. ID is 5-1⁄2" (14.0 cm). For your current glove box, order Iris diaphragm retrofit kit (34750-62).
Catalog No. NC1564217 Supplier Cole-Parmer Supplier No. 3475060
Must be ordered when glove box is ordered. ID is 5-1⁄2" (14.0 cm). For your current glove box, order Iris diaphragm retrofit kit (34750-62).