Learn More
Medicus Health Glove Dispensers, Quad Side Load Powder Coated Metal
SDP
Catalog No. NC3199660
Encompass
Side Load Powder Coated Metal Glove Dispensers have innovative side-load design that creates three mounting possibilities ∙ Unique keyhole design allows unit to be mounted with opening to the left, to the right or to the top.∙ Side Load Powder Coated Metal Glove Dispensers are made from 22 gauge powder coated steel to provide many years of service∙ Integral dividers to keep glove boxes separated and make glove box exchanges easier∙ Easy to clean with any typical hospital cleaner, even a 10% bleach solution.∙ Can be mounted on wall to save valuable counter space.∙ Quad Glove Dispenser dimensions are: 10"W x 4"D x 22"H
Catalog No. NC3199660 Supplier Medicus Health Supplier No. 2327M4
Side Load Powder Coated Metal Glove Dispensers have innovative side-load design that creates three mounting possibilities ∙ Unique keyhole design allows unit to be mounted with opening to the left, to the right or to the top.∙ Side Load Powder Coated Metal Glove Dispensers are made from 22 gauge powder coated steel to provide many years of service∙ Integral dividers to keep glove boxes separated and make glove box exchanges easier∙ Easy to clean with any typical hospital cleaner, even a 10% bleach solution.∙ Can be mounted on wall to save valuable counter space.∙ Quad Glove Dispenser dimensions are: 10"W x 4"D x 22"H