Learn More
Med Vet International McKesson Confiderm Latex Exam Glove, Large, Ivory, NonSterile, 14-1383, 100/Box
Catalog No. 503044078
Encompass_Preferred
Exam Glove McKesson Confiderm Large NonSterile Latex Standard Cuff Length Fully Textured Ivory Not Rated McKesson Confiderm gloves made of latex to reduce wear for continued reliable performance The ivory gloves are textured for ease of use and grasping items The standard beaded cuff prevents roll-down The powder-free large latex exam gloves have low protein content, helping to minimize allergic reactions Thin construction allows for improved tactile sensitivity The interior coating is non-sticking for activewear in wet, damp or dry situations
Catalog No. 50-304-4078 Supplier Med Vet International Supplier No. 921599BX
Exam Glove McKesson Confiderm Large NonSterile Latex Standard Cuff Length Fully Textured Ivory Not Rated McKesson Confiderm gloves made of latex to reduce wear for continued reliable performance The ivory gloves are textured for ease of use and grasping items The standard beaded cuff prevents roll-down The powder-free large latex exam gloves have low protein content, helping to minimize allergic reactions Thin construction allows for improved tactile sensitivity The interior coating is non-sticking for activewear in wet, damp or dry situations