Learn More
Med Vet International Exam Glove Polymed Large NonSterile Latex Standard Cuff Length Fully Textured Ivory Not Rated, PM104, 100/Box
Catalog No. 503043014
Encompass_Preferred
Eliminates the potential aerosolization of glove powder and powder-associated transfer of latex proteins Multi-layer, fusion bonded 50 micrograms or less of total water-extractable proteins per gram reduced potential for allergic reaction Micro-rough textured surface
Catalog No. 50-304-3014 Supplier Med Vet International Supplier No. 349006BX
Eliminates the potential aerosolization of glove powder and powder-associated transfer of latex proteins Multi-layer, fusion bonded 50 micrograms or less of total water-extractable proteins per gram reduced potential for allergic reaction Micro-rough textured surface