Learn More
Med Vet International McKesson Confiderm Latex Exam Glove, Large, Ivory, NonSterile, 14-428, 1000/Case
Catalog No. 503045094
Encompass_Preferred
Exam Glove McKesson Confiderm Large NonSterile Latex Standard Cuff Length Textured Fingertips Ivory Not Rated These large McKesson exam gloves are soft yet durable for continued reliable performance The slip-resistant, textured fingertips and fitted design let you pick up, use and retain objects in wet and dry conditions These ivory McKesson Confiderm gloves have a standard cuff length and a beaded cuff to reduce roll-down and avoid drips to the forearm The powder-free construction eliminates hand residue to protect against contamination The strong latex exam gloves reduce tears and punctures
Catalog No. 50-304-5094 Supplier Med Vet International Supplier No. 921594CS
Exam Glove McKesson Confiderm Large NonSterile Latex Standard Cuff Length Textured Fingertips Ivory Not Rated These large McKesson exam gloves are soft yet durable for continued reliable performance The slip-resistant, textured fingertips and fitted design let you pick up, use and retain objects in wet and dry conditions These ivory McKesson Confiderm gloves have a standard cuff length and a beaded cuff to reduce roll-down and avoid drips to the forearm The powder-free construction eliminates hand residue to protect against contamination The strong latex exam gloves reduce tears and punctures