Learn More
Med Vet International McKesson Confiderm Latex Exam Glove, Medium, Ivory, NonSterile, 14-1382, 100/Box
Catalog No. 503045125
Encompass_Preferred
Exam Glove McKesson Confiderm Medium NonSterile Latex Standard Cuff Length Fully Textured Ivory Not Rated McKesson exam gloves made of latex reduce wear for continued reliable performance The ivory gloves come textured on the outside for grasping items and ease of use The standard beaded cuff offers protection prevents roll-down The powder-free exam gloves provide a low protein content, helping with minimizing allergic reactions The medium latex exam gloves have an interior coating for anti-sticking wear in damp, wet or dry conditions They are not chemo approved
Catalog No. 50-304-5125 Supplier Med Vet International Supplier No. 921598BX
Add to Cart
Add to Cart
Exam Glove McKesson Confiderm Medium NonSterile Latex Standard Cuff Length Fully Textured Ivory Not Rated McKesson exam gloves made of latex reduce wear for continued reliable performance The ivory gloves come textured on the outside for grasping items and ease of use The standard beaded cuff offers protection prevents roll-down The powder-free exam gloves provide a low protein content, helping with minimizing allergic reactions The medium latex exam gloves have an interior coating for anti-sticking wear in damp, wet or dry conditions They are not chemo approved