Learn More
Med Vet International Exam Glove Polymed Medium NonSterile Latex Standard Cuff Length Fully Textured Ivory Not Rated, PM103, 100/Box
Supplier:  Med Vet International 349005BX
Encompass_Preferred
Eliminates the potential aerosolization of glove powder and powder-associated transfer of latex proteins Multi-layer, fusion bonded 50 micrograms or less of total water-extractable proteins per gram reduced potential for allergic reaction Micro-rough textured surface
Catalog No. 50-304-3537