Learn More
Med Vet International McKesson Confiderm Latex Exam Glove, Medium, Ivory, NonSterile, 14-316, 100/Box
Catalog No. 503043018
Encompass_Preferred
Exam Glove McKesson Confiderm Medium NonSterile Latex Standard Cuff Length Smooth Ivory Not Rated McKesson exam gloves' smooth surface, inside and out, make for convenient on and off access They're durable composition naturally resists tears and punctures for effective, reliable use The ivory, latex exam gloves have a standard cuff and a beaded design that prevents roll-down and reduces drips to the forearm Made without powder, they won't leave behind any residue, reducing the risk of cross-contamination The size small gloves are ambidextrous, suitable for use on the left or right hand
Catalog No. 50-304-3018 Supplier Med Vet International Supplier No. 354436BX
Exam Glove McKesson Confiderm Medium NonSterile Latex Standard Cuff Length Smooth Ivory Not Rated McKesson exam gloves' smooth surface, inside and out, make for convenient on and off access They're durable composition naturally resists tears and punctures for effective, reliable use The ivory, latex exam gloves have a standard cuff and a beaded design that prevents roll-down and reduces drips to the forearm Made without powder, they won't leave behind any residue, reducing the risk of cross-contamination The size small gloves are ambidextrous, suitable for use on the left or right hand