Learn More
Med Vet International McKesson Confiderm Latex Exam Glove, Small, Ivory, NonSterile, 14-1381, 1000/Case
Catalog No. 503045211
Encompass_Preferred
Exam Glove McKesson Confiderm Small NonSterile Latex Standard Cuff Length Fully Textured Ivory Not Rated McKesson exam gloves made of latex reduce wear for continued reliable performance The surface is smooth on the inside with a polymer coating allows for ease of use The standard beaded cuff offers protection beyond the wrist and prevents roll-down The powder-free, small latex exam gloves lower the risk of allergic reactions Thin construction and textured fingertips allows for improved tactile sensitivity The ivory McKesson Confiderm gloves are ambidextrous permitting use on either hand
Catalog No. 50-304-5211 Supplier Med Vet International Supplier No. 921597CS
Exam Glove McKesson Confiderm Small NonSterile Latex Standard Cuff Length Fully Textured Ivory Not Rated McKesson exam gloves made of latex reduce wear for continued reliable performance The surface is smooth on the inside with a polymer coating allows for ease of use The standard beaded cuff offers protection beyond the wrist and prevents roll-down The powder-free, small latex exam gloves lower the risk of allergic reactions Thin construction and textured fingertips allows for improved tactile sensitivity The ivory McKesson Confiderm gloves are ambidextrous permitting use on either hand