Learn More
Med Vet International McKesson Confiderm Latex Exam Glove, Small, Ivory, NonSterile, 14-424, 1000/Case
Catalog No. 503044105
Encompass_Preferred
Exam Glove McKesson Confiderm Small NonSterile Latex Standard Cuff Length Textured Fingertips Ivory Not Rated These small exam gloves are soft yet durable for continued reliable performance The slip-resistant, textured fingertips let you pick up, use and retain objects in wet and dry conditions The ivory-colored McKesson Confiderm gloves have a standard cuff length and a beaded cuff to reduce roll-down and avoid drips to the forearm The powder-free construction eliminates hand residue to protect against contamination The strong latex rubber reduces tears and punctures
Catalog No. 50-304-4105 Supplier Med Vet International Supplier No. 921592CS
Exam Glove McKesson Confiderm Small NonSterile Latex Standard Cuff Length Textured Fingertips Ivory Not Rated These small exam gloves are soft yet durable for continued reliable performance The slip-resistant, textured fingertips let you pick up, use and retain objects in wet and dry conditions The ivory-colored McKesson Confiderm gloves have a standard cuff length and a beaded cuff to reduce roll-down and avoid drips to the forearm The powder-free construction eliminates hand residue to protect against contamination The strong latex rubber reduces tears and punctures