Learn More
Med Vet International McKesson Confiderm Latex Exam Glove, Small, Ivory, NonSterile, 14-314, 1000/Case
Catalog No. 503044971
Encompass_Preferred
Exam Glove McKesson Confiderm Small NonSterile Latex Standard Cuff Length Smooth Ivory Not Rated McKesson Confiderm gloves feature a smooth surface inside and out for convenient on and off access They're durable composition naturally resists tears and punctures for effective, reliable use The ivory latex exam gloves have a standard cuff and a beaded design that prevents roll-down and reduces drips to the forearm Made without powder, they won't leave behind any residue, reducing the risk of cross-contamination The size small gloves are ambidextrous, suitable for use on the left or right hand
Catalog No. 50-304-4971 Supplier Med Vet International Supplier No. 354435CS
Exam Glove McKesson Confiderm Small NonSterile Latex Standard Cuff Length Smooth Ivory Not Rated McKesson Confiderm gloves feature a smooth surface inside and out for convenient on and off access They're durable composition naturally resists tears and punctures for effective, reliable use The ivory latex exam gloves have a standard cuff and a beaded design that prevents roll-down and reduces drips to the forearm Made without powder, they won't leave behind any residue, reducing the risk of cross-contamination The size small gloves are ambidextrous, suitable for use on the left or right hand