Learn More
Med Vet International McKesson Confiderm Latex Exam Glove, Extra Large, Ivory, NonSterile, 14-1384, 100/Box
Catalog No. 503043098
Encompass_Preferred
Exam Glove McKesson Confiderm X-Large NonSterile Latex Standard Cuff Length Fully Textured Ivory Not Rated McKesson exam gloves made of latex reduce wear for continued reliable performance The surface is smooth on the inside featuring a polymer coating for ease of use The standard beaded cuff offers protection beyond the wrist and prevent roll-down The ivory, powder-free exam gloves lower the risk of allergic reactions and cross contamination The thin construction and textured exterior allows for improved tactile sensitivity The ambidextrous construction permits use on the left and right hand
Catalog No. 50-304-3098 Supplier Med Vet International Supplier No. 921600BX
Exam Glove McKesson Confiderm X-Large NonSterile Latex Standard Cuff Length Fully Textured Ivory Not Rated McKesson exam gloves made of latex reduce wear for continued reliable performance The surface is smooth on the inside featuring a polymer coating for ease of use The standard beaded cuff offers protection beyond the wrist and prevent roll-down The ivory, powder-free exam gloves lower the risk of allergic reactions and cross contamination The thin construction and textured exterior allows for improved tactile sensitivity The ambidextrous construction permits use on the left and right hand