Learn More
Med Vet International McKesson Confiderm Latex Exam Glove, Extra Large, Ivory, NonSterile, 14-430, 1000/Case
Catalog No. 503045142
Encompass_Preferred
Exam Glove McKesson Confiderm X-Large NonSterile Latex Standard Cuff Length Textured Fingertips Ivory Not Rated McKesson exam gloves are soft yet durable for continued reliable performance The slip-resistant, textured fingertips and fitted design let you pick up, use and retain objects in wet and dry conditions The extra-large McKesson Confiderm ivory gloves have a standard cuff length and a beaded cuff to reduce roll-down The powder-free construction eliminates hand residue to protect against contamination The strong latex rubber reduces tears and punctures The ambidextrous construction permits use on the left and right hand
Catalog No. 50-304-5142 Supplier Med Vet International Supplier No. 921595CS
Exam Glove McKesson Confiderm X-Large NonSterile Latex Standard Cuff Length Textured Fingertips Ivory Not Rated McKesson exam gloves are soft yet durable for continued reliable performance The slip-resistant, textured fingertips and fitted design let you pick up, use and retain objects in wet and dry conditions The extra-large McKesson Confiderm ivory gloves have a standard cuff length and a beaded cuff to reduce roll-down The powder-free construction eliminates hand residue to protect against contamination The strong latex rubber reduces tears and punctures The ambidextrous construction permits use on the left and right hand