Learn More
Med Vet International McKesson Confiderm Latex Exam Glove, Extra Large, Ivory, NonSterile, 14-320, 1000/Case
Catalog No. 503044446
Encompass_Preferred
Exam Glove McKesson Confiderm X-Large NonSterile Latex Standard Cuff Length Smooth Ivory Not Rated McKesson exam gloves' smooth surface, inside and out, make for convenient on and off access They're durable composition naturally resists tears and punctures for effective, reliable use The x-large latex exam gloves have a standard cuff and a beaded design that prevents roll-down and reduces drips to the forearm Made without powder, they won't leave behind any residue, reducing the risk of cross-contamination The ivory gloves are ambidextrous, suitable for use on the left or right hand
Catalog No. 50-304-4446 Supplier Med Vet International Supplier No. 483972CS
Exam Glove McKesson Confiderm X-Large NonSterile Latex Standard Cuff Length Smooth Ivory Not Rated McKesson exam gloves' smooth surface, inside and out, make for convenient on and off access They're durable composition naturally resists tears and punctures for effective, reliable use The x-large latex exam gloves have a standard cuff and a beaded design that prevents roll-down and reduces drips to the forearm Made without powder, they won't leave behind any residue, reducing the risk of cross-contamination The ivory gloves are ambidextrous, suitable for use on the left or right hand