Learn More
Med Vet International McKesson Extra Small Non-Sterile, Latex, Standard Cuff Length, Not Chemo Approved Exam Gloves, Ivory, 14-1380, 1000/Case
Catalog No. 503045096
Encompass_Preferred
Exam Glove McKesson Confiderm X-Small NonSterile Latex Standard Cuff Length Fully Textured Ivory Not Rated McKesson exam gloves made of vinyl reduce wear for continued reliable performance The surface of these x-small latex exam gloves is smooth on the inside and outside for ease of use The standard beaded cuff offers protection beyond the wrist and prevents roll-down The powder-free ivory exam gloves have low protein content lowering the risk of allergic reactions Thin construction allows for improved tactile sensitivity
Catalog No. 50-304-5096 Supplier Med Vet International Supplier No. 921596CS
Exam Glove McKesson Confiderm X-Small NonSterile Latex Standard Cuff Length Fully Textured Ivory Not Rated McKesson exam gloves made of vinyl reduce wear for continued reliable performance The surface of these x-small latex exam gloves is smooth on the inside and outside for ease of use The standard beaded cuff offers protection beyond the wrist and prevents roll-down The powder-free ivory exam gloves have low protein content lowering the risk of allergic reactions Thin construction allows for improved tactile sensitivity