Learn More
Med Vet International McKesson Confiderm CL Latex Gloves, Extra-Small, Ivory, NonSterile, 14-422, 1000/Case
Supplier:  Med Vet International 921591CS
Encompass_Preferred
Exam Glove McKesson Confiderm CL X-Small NonSterile Latex Standard Cuff Length Textured Fingertips Ivory Not Rated Made of latex for excellent tactile sensitivity Like all medical gloves, these are powder free Textured fingertips give slip-resistant precision when performing intricate tasks in wet or dry conditions Beaded cuffs keep the gloves secure and provide easy on and off Size extra-small (ambidextrous) Extra small
Catalog No. 50-304-5140