Learn More
Med Vet International Glove Exam Nitrile Lg Pf Purple 1000 Ea/Case
Catalog No. 502831756
Encompass_Preferred
Disposable Nitrile Exam Gloves, Large, Purple, 100 per Box. Get a good fit and a good grip. The textured fingertips on these disposable gloves will allow you to work with ease and confidence. Beaded cuffs provide easier donning and added strength. No natural rubber latex to cause possible allergic reaction. Powder-free. Ambidextrous. 9 1/2" long.
Catalog No. 50-283-1756 Supplier Med Vet International Supplier No. KC55083
Disposable Nitrile Exam Gloves, Large, Purple, 100 per Box. Get a good fit and a good grip. The textured fingertips on these disposable gloves will allow you to work with ease and confidence. Beaded cuffs provide easier donning and added strength. No natural rubber latex to cause possible allergic reaction. Powder-free. Ambidextrous. 9 1/2" long.