Learn More
Med Vet International Purple Nitrile Exam Glove, Medium, NonSterile, Nitrile, Standard Cuff Length, Textured Fingertips, Chemo Tested, 55082, 1000/Case
Catalog No. 503044593
Encompass_Preferred
Purple Nitrile exam gloves are effective against chemicals, bacteria and viruses in medium- to high-risk environments They're effective for protection during chemotherapy procedures Like all medical exam gloves, these medium Purple Nitrile gloves are free of powder 9½ inches in length Textured fingertips offer a better grip in both wet and dry conditions They exceed AQL requirements with a rating of 1.0
Catalog No. 50-304-4593 Supplier Med Vet International Supplier No. 365061CS
Purple Nitrile exam gloves are effective against chemicals, bacteria and viruses in medium- to high-risk environments They're effective for protection during chemotherapy procedures Like all medical exam gloves, these medium Purple Nitrile gloves are free of powder 9½ inches in length Textured fingertips offer a better grip in both wet and dry conditions They exceed AQL requirements with a rating of 1.0