Learn More
Med Vet International MICROFLEX MidKnight Touch 93-733 Exam Gloves, Small, NonSterile, Nitrile, Standard Cuff Length, Textured Fingertips, Black, 93732070, 100/Box
Catalog No. 503044940
Encompass_Preferred
3.9 mil fingertip thickness Textured fingertips Standard-length beaded cuff Ambidextrous 9-2/3-inch length Not tested for chemo Small size is for those whose palm circumference is between 6½ and 7 inches
Catalog No. 50-304-4940 Supplier Med Vet International Supplier No. 1182922BX
3.9 mil fingertip thickness Textured fingertips Standard-length beaded cuff Ambidextrous 9-2/3-inch length Not tested for chemo Small size is for those whose palm circumference is between 6½ and 7 inches