Learn More
Med Vet International McKesson Confiderm 3.0 Nitrile Exam Glove, Large, Blue, 14-6N36EC, 1000/Case
Catalog No. 503042942
Encompass_Preferred
Exam Glove McKesson Confiderm 3.0 Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated Textured fingertips give these nonsterile gloves outstanding tactile sensitivity Thin texture and high elasticity provide a comfortable but snug conforming fit These large blue nitrile gloves are ambidextrous for convenient wear Beaded cuffs prevent them from rolling up
Catalog No. 50-304-2942 Supplier Med Vet International Supplier No. 1107942CS
Exam Glove McKesson Confiderm 3.0 Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated Textured fingertips give these nonsterile gloves outstanding tactile sensitivity Thin texture and high elasticity provide a comfortable but snug conforming fit These large blue nitrile gloves are ambidextrous for convenient wear Beaded cuffs prevent them from rolling up