Learn More
Med Vet International McKesson Confiderm 3.5C Nitrile Exam Glove, Large, Blue, 14-6978C, 200/Box
Catalog No. 503044366
Encompass_Preferred
Exam Glove McKesson Confiderm 3.5C Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Chemo Tested McKesson Confiderm gloves are single-use and disposable to maintain a hygienic clinical environment They are ambidextrous to ensure a quick and easy fit on either hand Confiderm 3.5C blue exam gloves are powder- and latex-free for reduced chemical sensitivity Textured fingertips increase sensitivity, grip and dexterity The standard-length beaded cuff forms a closed seal around the wrist and provides easy removal These nitrile exam gloves meet requirements for handling chemotherapy drugs Large
Catalog No. 50-304-4366 Supplier Med Vet International Supplier No. 765876BX
Exam Glove McKesson Confiderm 3.5C Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Chemo Tested McKesson Confiderm gloves are single-use and disposable to maintain a hygienic clinical environment They are ambidextrous to ensure a quick and easy fit on either hand Confiderm 3.5C blue exam gloves are powder- and latex-free for reduced chemical sensitivity Textured fingertips increase sensitivity, grip and dexterity The standard-length beaded cuff forms a closed seal around the wrist and provides easy removal These nitrile exam gloves meet requirements for handling chemotherapy drugs Large