Learn More
Med Vet International McKesson Confiderm 3.8 Nitrile Exam Glove, Large, Blue, NonSterile, 14-688, 1000/Case
Catalog No. 503045071
Encompass_Preferred
Exam Glove McKesson Confiderm 3.8 Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated The textured fingertips offer enhanced grip and finger movement Beaded cuffs help keep the gloves in place These blue nitrile exam gloves are not made with natural rubber latex to reduce irritation The gloves are wearable on left or right hand Confiderm large exam gloves conform well to hand shapes to keep user comfortable They have a standard cuff length and are not chemo approved
Catalog No. 50-304-5071 Supplier Med Vet International Supplier No. 921613CS
Exam Glove McKesson Confiderm 3.8 Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated The textured fingertips offer enhanced grip and finger movement Beaded cuffs help keep the gloves in place These blue nitrile exam gloves are not made with natural rubber latex to reduce irritation The gloves are wearable on left or right hand Confiderm large exam gloves conform well to hand shapes to keep user comfortable They have a standard cuff length and are not chemo approved