Learn More
Med Vet International Exam Glove Purple Nitrile-Xtra™ Large NonSterile Nitrile Extended Cuff Length Textured Fingertips Purple Chemo Tested, 50603, 50/Box
Catalog No. 503042930
Encompass_Preferred
Halyard Purple Nitrile-Xtra exam gloves have textured fingertips Their unique purple color makes it easy to locate them They are long for maximum coverage and protection These large purple Halyard Nitrile-Xtra gloves have an enhanced grip for support on wet areas They're extra thick to protect against high and moderate exposure to chemicals, bacteria and fluids
Catalog No. 50-304-2930 Supplier Med Vet International Supplier No. 365067BX
Halyard Purple Nitrile-Xtra exam gloves have textured fingertips Their unique purple color makes it easy to locate them They are long for maximum coverage and protection These large purple Halyard Nitrile-Xtra gloves have an enhanced grip for support on wet areas They're extra thick to protect against high and moderate exposure to chemicals, bacteria and fluids