Learn More
Med Vet International Exam Glove MICROFLEX MidKnight™ Touch 93-735 Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Black Not Rated, 93732090, 1000/Case
Catalog No. 503044052
Encompass_Preferred
3.9 mil fingertip thickness Textured fingertips Standard-length beaded cuff Ambidextrous 9?-inch length Large size is for those whose palm circumference is between 8½ and 9 inches
Catalog No. 50-304-4052 Supplier Med Vet International Supplier No. 1182924CS
3.9 mil fingertip thickness Textured fingertips Standard-length beaded cuff Ambidextrous 9?-inch length Large size is for those whose palm circumference is between 8½ and 9 inches