Learn More
Med Vet International Exam Glove Purple Nitrile Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Purple Chemo Tested, 55083, 100/Box
Catalog No. 503044710
Encompass_Preferred
Purple Nitrile exam gloves are effective against chemicals, bacteria and viruses in medium- to high-risk environments They're effective for protection during chemotherapy procedures Like all medical exam gloves, these large Purple Nitrile gloves are free of powder 9½ inches in length Textured fingertips offer a better grip in both wet and dry conditions They exceed AQL requirements with a rating of 1.0
Catalog No. 50-304-4710 Supplier Med Vet International Supplier No. 365062BX
Purple Nitrile exam gloves are effective against chemicals, bacteria and viruses in medium- to high-risk environments They're effective for protection during chemotherapy procedures Like all medical exam gloves, these large Purple Nitrile gloves are free of powder 9½ inches in length Textured fingertips offer a better grip in both wet and dry conditions They exceed AQL requirements with a rating of 1.0