Learn More
Med Vet International Exam Glove STERLING Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Gray Chemo Tested, 50708, 200/Box
Catalog No. 503044296
Encompass_Preferred
Nitrile material allows for comfortable wear Textured fingertips provide reliable grip Beaded cuffs help keep the gloves in place Chemo tested (Cyclophosphamide, Doxorubicin HCl, Etoposide, 5-Fluorouracil, Paclitaxel (Taxol), Cisplatin, Decarbonize, Ifosfamide, Mitoxantrone, Vincristine Sulfate) Nonsterile and designed for one-time use Like all medical gloves, these are powder free
Catalog No. 50-304-4296 Supplier Med Vet International Supplier No. 761282BX
Nitrile material allows for comfortable wear Textured fingertips provide reliable grip Beaded cuffs help keep the gloves in place Chemo tested (Cyclophosphamide, Doxorubicin HCl, Etoposide, 5-Fluorouracil, Paclitaxel (Taxol), Cisplatin, Decarbonize, Ifosfamide, Mitoxantrone, Vincristine Sulfate) Nonsterile and designed for one-time use Like all medical gloves, these are powder free