Learn More
Med Vet International Exam Glove PremierPro™ Plus Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Chemo Tested, 5064, 200/Box
Catalog No. 503045143
Encompass_Preferred
Fingertip textured; fingertip thickness is 0.09 mm (3.5 mil) Complies with standard ASTM 6978 (Assessment of Medical Exam Gloves for Resistance to Permeation by Chemotherapy Drugs) Meets USP 800 Personal Protection Equipment requirement for use of Chemotherapy Gloves when handling Hazardous Drugs Nonsterile Beaded cuff Ambidextrous Like all medical exam gloves, these are powder free Large
Catalog No. 50-304-5143 Supplier Med Vet International Supplier No. 897351BX
Fingertip textured; fingertip thickness is 0.09 mm (3.5 mil) Complies with standard ASTM 6978 (Assessment of Medical Exam Gloves for Resistance to Permeation by Chemotherapy Drugs) Meets USP 800 Personal Protection Equipment requirement for use of Chemotherapy Gloves when handling Hazardous Drugs Nonsterile Beaded cuff Ambidextrous Like all medical exam gloves, these are powder free Large