Learn More
Med Vet International Exam Glove STERLING Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Gray Chemo Tested, 50708, 2000/Case
Catalog No. 503044297
Encompass_Preferred
Nitrile material allows for comfortable wear Textured fingertips provide reliable grip Beaded cuffs help keep the gloves in place Chemo tested (Cyclophosphamide, Doxorubicin HCl, Etoposide, 5-Fluorouracil, Paclitaxel (Taxol), Cisplatin, Decarbonize, Ifosfamide, Mitoxantrone, Vincristine Sulfate) Nonsterile and designed for one-time use Like all medical gloves, these are powder free
Catalog No. 50-304-4297 Supplier Med Vet International Supplier No. 761282CS
Nitrile material allows for comfortable wear Textured fingertips provide reliable grip Beaded cuffs help keep the gloves in place Chemo tested (Cyclophosphamide, Doxorubicin HCl, Etoposide, 5-Fluorouracil, Paclitaxel (Taxol), Cisplatin, Decarbonize, Ifosfamide, Mitoxantrone, Vincristine Sulfate) Nonsterile and designed for one-time use Like all medical gloves, these are powder free