Learn More
Med Vet International Exam Glove Pulse Nitrile Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Aqua Blue Chemo Tested, 177302, 2000/Case
Catalog No. 503044283
Encompass_Preferred
Pulse Nitrile gloves have been tested for use with chemotherapy drugs Finely textured finger tips create an unequalled tactile sensitivity and excellent gripping properties Large
Catalog No. 50-304-4283 Supplier Med Vet International Supplier No. 764271CS
Pulse Nitrile gloves have been tested for use with chemotherapy drugs Finely textured finger tips create an unequalled tactile sensitivity and excellent gripping properties Large