Learn More
Med Vet International Exam Glove Micro-Touch Nitrile Large NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Chemo Tested, 6034303, 2000/Case
Catalog No. 503043284
Encompass_Preferred
They offer a snug, comfortable fit for better tactile sensitivity The nonsterile gloves are sturdy and durable Tested for use with 55 chemicals, and approved for chemotherapy drugs The Micro-Touch gloves are not made from natural rubber latex and will not irritate the skin The gloves meet or exceed industry standards for consistent production quality Large size is for those whose palm circumference is between 8½ and 9 inches
Catalog No. 50-304-3284 Supplier Med Vet International Supplier No. 703152CS
They offer a snug, comfortable fit for better tactile sensitivity The nonsterile gloves are sturdy and durable Tested for use with 55 chemicals, and approved for chemotherapy drugs The Micro-Touch gloves are not made from natural rubber latex and will not irritate the skin The gloves meet or exceed industry standards for consistent production quality Large size is for those whose palm circumference is between 8½ and 9 inches