Learn More
Med Vet International Exam Glove Purple Nitrile Large Sterile Pair Nitrile Standard Cuff Length Textured Fingertips Purple Not Rated, 55093, Pair
Catalog No. 503044850
Encompass_Preferred
Like all medical exam gloves, these are powder free The gloves meet ASTM requirements, highlighting their reliability Halyard Purple Nitrile exam gloves have textured fingertips to provide an improved grip The beaded cuff style adds better fit and extra protection These gloves are packed in sterile pairs Large
Catalog No. 50-304-4850 Supplier Med Vet International Supplier No. 407604EA
Like all medical exam gloves, these are powder free The gloves meet ASTM requirements, highlighting their reliability Halyard Purple Nitrile exam gloves have textured fingertips to provide an improved grip The beaded cuff style adds better fit and extra protection These gloves are packed in sterile pairs Large