Learn More
Med Vet International McKesson Confiderm STR Nitrile Exam Glove, Large, Blue, 14-6NSTR6, Pair
Catalog No. 503043735
Encompass_Preferred
Exam Glove McKesson Confiderm STR Large Sterile Pair Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated These large nitrile exam gloves resist tear and punctures for reliable performance Beaded cuffs reduce drips and prevent roll-up The textured fingertips improve tactile sensitivity and agility Powder-free construction eliminates hand residue The McKesson nitrile exam gloves offer a superior fit to maintain comfort They're latex-free, making them safe for those with latex allergies
Catalog No. 50-304-3735 Supplier Med Vet International Supplier No. 1065407PR
Exam Glove McKesson Confiderm STR Large Sterile Pair Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated These large nitrile exam gloves resist tear and punctures for reliable performance Beaded cuffs reduce drips and prevent roll-up The textured fingertips improve tactile sensitivity and agility Powder-free construction eliminates hand residue The McKesson nitrile exam gloves offer a superior fit to maintain comfort They're latex-free, making them safe for those with latex allergies