Learn More
Med Vet International Exam Glove Micro-Touch NitraFree™ Medium NonSterile Nitrile Standard Cuff Length Textured Fingertips Pink Chemo Tested, 6034512, 100/Box
Catalog No. 503043718
Encompass_Preferred
Ansell's Micro-Touch NitraFree exam gloves protect against latex Type I allergies and reduce chemical Type IV allergies They're ideal for performing exams or caring for ill family members at home These lightweight gloves offer a secure fit and feature textured fingers and a standard cuff length Use these gloves for protection against harsh chemicals and cleaners These powder-free exam gloves are nonsterile and latex-free This disposable product is safe to use with chemotherapy medication Medium size is for those whose palm circumference is between 7½ and 8 inches
Catalog No. 50-304-3718 Supplier Med Vet International Supplier No. 693951BX
Ansell's Micro-Touch NitraFree exam gloves protect against latex Type I allergies and reduce chemical Type IV allergies They're ideal for performing exams or caring for ill family members at home These lightweight gloves offer a secure fit and feature textured fingers and a standard cuff length Use these gloves for protection against harsh chemicals and cleaners These powder-free exam gloves are nonsterile and latex-free This disposable product is safe to use with chemotherapy medication Medium size is for those whose palm circumference is between 7½ and 8 inches