Learn More
Med Vet International McKesson Confiderm 4.5C Nitrile Exam Glove, Medium, Blue, NonSterile, 14-6N34, 2500/Case
Catalog No. 503045388
Encompass_Preferred
Exam Glove McKesson Confiderm 3.0 Medium NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated Textured fingertips offer outstanding tactile sensitivity Confiderm nitrile exam gloves have beaded cuffs to prevent them from rolling up Their outstanding conformity offers a proper fit These powder-free, blue, medium nitrile exam gloves are wearable on left or right hand
Catalog No. 50-304-5388 Supplier Med Vet International Supplier No. 957802CS
Exam Glove McKesson Confiderm 3.0 Medium NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated Textured fingertips offer outstanding tactile sensitivity Confiderm nitrile exam gloves have beaded cuffs to prevent them from rolling up Their outstanding conformity offers a proper fit These powder-free, blue, medium nitrile exam gloves are wearable on left or right hand