Learn More
Med Vet International McKesson Confiderm 3.8 Nitrile Exam Glove, Medium, Blue, NonSterile, 14-686, 1000/Case
Supplier:  Med Vet International 921611CS
Encompass_Preferred
Exam Glove McKesson Confiderm 3.8 Medium NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Not Rated The textured fingertips offer enhanced grip and finger movement Beaded cuffs help keep gloves in place These blue nitrile exam gloves are not made with natural rubber latex to reduce irritation They are wearable on the left or right hand Confiderm medium exam gloves conform well to hand shapes to keep user comfortable Medium
Catalog No. 50-304-5166