Learn More
Med Vet International Exam Glove Micro-Touch Nitrile Medium NonSterile Nitrile Standard Cuff Length Textured Fingertips Blue Chemo Tested, 6034302, 2000/Case
Catalog No. 503044879
Encompass_Preferred
The Ansell Micro-Touch nitrile gloves offer a snug, comfortable fit for better tactile sensitivity The blue, medium size nonsterile gloves are sturdy and durable They're tested for use with 55 chemicals, and approved for chemotherapy drugs The Micro-Touch gloves are not made from natural rubber latex and will not irritate the skin The gloves meet or exceed industry standards for consistent production quality Medium size is for those whose palm circumference is between 7½ and 8 inches
Catalog No. 50-304-4879 Supplier Med Vet International Supplier No. 703151CS
The Ansell Micro-Touch nitrile gloves offer a snug, comfortable fit for better tactile sensitivity The blue, medium size nonsterile gloves are sturdy and durable They're tested for use with 55 chemicals, and approved for chemotherapy drugs The Micro-Touch gloves are not made from natural rubber latex and will not irritate the skin The gloves meet or exceed industry standards for consistent production quality Medium size is for those whose palm circumference is between 7½ and 8 inches