Learn More
Med Vet International Exam Glove Purple Nitrile-Xtra™ Medium Sterile Pair Nitrile Extended Cuff Length Textured Fingertips Purple Chemo Tested, 14261, 200 Pairs
Catalog No. 503043413
Encompass_Preferred
These medium nitrile exam gloves provide maximum coverage Beaded cuff prevents roll-down and drips They're powder-free to reduce the risk of cross-contamination These Halyard Purple Nitrile exam gloves have textured fingertips that improve tactile sensitivity They have a latex-free construction that's suitable for all skin types These nitrile powder-free exam gloves are made of quality materials to prevent contact with fluids and chemicals
Catalog No. 50-304-3413 Supplier Med Vet International Supplier No. 1042398CS
These medium nitrile exam gloves provide maximum coverage Beaded cuff prevents roll-down and drips They're powder-free to reduce the risk of cross-contamination These Halyard Purple Nitrile exam gloves have textured fingertips that improve tactile sensitivity They have a latex-free construction that's suitable for all skin types These nitrile powder-free exam gloves are made of quality materials to prevent contact with fluids and chemicals