Learn More
Med Vet International Exam Glove Purple Nitrile Medium Sterile Pair Nitrile Standard Cuff Length Textured Fingertips Purple Not Rated, 55092, 200 Pairs
Catalog No. 503042851
Encompass_Preferred
Like all medical exam gloves, these are powder free The gloves meet ASTM requirements, highlighting their reliability Halyard Purple Nitrile exam gloves have textured fingertips to provide an improved grip The beaded cuff style adds better fit and extra protection These gloves are packed in sterile pairs Medium
Catalog No. 50-304-2851 Supplier Med Vet International Supplier No. 407603CS
Like all medical exam gloves, these are powder free The gloves meet ASTM requirements, highlighting their reliability Halyard Purple Nitrile exam gloves have textured fingertips to provide an improved grip The beaded cuff style adds better fit and extra protection These gloves are packed in sterile pairs Medium